Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129758.1 | NP_001123230.1 | 5 | 886 | ||
NM_022357.3 | NP_071752.3 | 5 | 886 | |||
GenBank mRNA | AJ291679.1 | - | 5 | 800 | ||
AK057401.1 | - | 5 | 886 | |||
AY358390.1 | - | 5 | 809 | |||
BC037243.1 | - | 5 | 798 | |||
BC057789.1 | - | 5 | 827 | 5297958 |