Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MMS19 SIRNA Assays ›
Gene Symbol:
MMS19
Gene Name:
MMS19 homolog, cytosolic iron-sulfur assembly component
Gene Aliases:
MET18, MMS19L, hMMS19
Chromosome Location:
Chr.10: 97458324 - 97498794 on Build GRCh38
UniGene ID:
Hs.500721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34552
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MMS19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289403.1
NP_001276332.1
24
2524
NM_001289404.1
NP_001276333.1
24
2589
NM_001289405.1
NP_001276334.1
26
2535
NM_022362.4
NP_071757.4
25
2653
XM_005270035.2
XP_005270092.1
24
2410
XM_005270041.1
XP_005270098.1
23
2298
XM_006717944.3
XP_006718007.2
25
2444
XM_006717945.2
XP_006718008.1
23
2986
XM_011540062.1
XP_011538364.1
24
2256
XM_011540063.2
XP_011538365.1
23
2984
XM_017016515.1
XP_016872004.1
26
2559
XM_017016516.1
XP_016872005.1
26
2556
XM_017016517.1
XP_016872006.1
25
2430
XM_017016518.1
XP_016872007.1
24
2315
XM_017016519.1
XP_016872008.1
24
2265
XM_017016520.1
XP_016872009.1
23
2153
XM_017016521.1
XP_016872010.1
23
2150
XM_017016522.1
XP_016872011.1
25
2551
XM_017016523.1
XP_016872012.1
24
2435
XM_017016524.1
XP_016872013.1
24
2407
XM_017016525.1
XP_016872014.1
23
2177
XM_017016526.1
XP_016872015.1
24
2413
XM_017016527.1
XP_016872016.1
24
2854
XM_017016528.1
XP_016872017.1
22
2825
XM_017016529.1
XP_016872018.1
24
3054
XM_017016530.1
XP_016872019.1
25
2573
XM_017016531.1
XP_016872020.1
23
2271
XM_017016532.1
XP_016872021.1
22
2048
XM_017016533.1
XP_016872022.1
22
2125
XM_017016534.1
XP_016872023.1
23
2981
XM_017016535.1
XP_016872024.1
22
2842
GenBank mRNA
AF007151.1
-
11
1115
AF319947.1
-
26
2501
AF357881.1
-
25
2452
AJ306408.1
-
25
2435
AK025496.1
-
12
1152
AK027710.1
-
22
1993
AK056244.1
-
14
1460
AK056581.1
-
19
1790
AK225312.1
-
25
2438
AK298995.1
-
25
2491
AK304287.1
-
24
2318
AK314374.1
-
25
2451
BC002692.1
-
14
1415
BC006575.2
-
17
1747
BC009396.2
-
8
719
BC080532.1
-
24
2368
4889876
BC117129.1
-
25
2451
40125680
BU942293.1
-
3
154
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
DNA metabolic process
DNA repair
nucleotide-excision repair
transcription, DNA-templated
cellular response to DNA damage stimulus
chromosome segregation
response to hormone
iron-sulfur cluster assembly
positive regulation of transcription, DNA-templated
transcription coactivator activity
protein binding
enzyme binding
receptor signaling complex scaffold activity
estrogen receptor binding
protein binding, bridging
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline