Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040284.2 | NP_001035374.2 | 3 | 720 | ||
NM_001040285.2 | NP_001035375.2 | 3 | 720 | |||
XM_005256092.4 | XP_005256149.1 | 2 | 903 | |||
XM_005256093.4 | XP_005256150.2 | 3 | 717 | |||
XM_011523275.2 | XP_011521577.1 | 3 | 716 | |||
XM_011523276.2 | XP_011521578.1 | 3 | 718 | |||
XM_011523278.2 | XP_011521580.1 | 2 | 191 | |||
XM_017023555.1 | XP_016879044.1 | 3 | 776 | |||
GenBank mRNA | AK097589.1 | - | 3 | 535 | ||
AK300918.1 | - | 3 | 720 | |||
DN994465.1 | - | 2 | 338 | |||
FR872509.1 | - | 2 | 484 |