Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080479.2 | NP_001073948.2 | 12 | 1747 | ||
NM_001177693.1 | NP_001171164.1 | 12 | 1747 | |||
NM_001244364.1 | NP_001231293.1 | 4 | 703 | |||
XM_005248568.4 | XP_005248625.1 | 4 | 2184 | |||
XM_011543546.1 | XP_011541848.1 | 12 | 1754 | |||
XM_011543547.2 | XP_011541849.1 | 11 | 1981 | |||
XM_017009702.1 | XP_016865191.1 | 11 | 1432 | |||
XM_017009703.1 | XP_016865192.1 | 3 | 485 | |||
GenBank mRNA | AB082529.1 | - | 11 | 1586 | ||
AK094713.1 | - | 12 | 1747 | |||
AK304761.1 | - | 4 | 703 | |||
BC157846.1 | - | 11 | 1593 | 9057065 | ||
BC171850.1 | - | 11 | 1593 | 9054504 |