Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135215.1 | NP_001128687.1 | 8 | 1661 | ||
NM_001135216.1 | NP_001128688.1 | 4 | 1157 | |||
NM_001243241.1 | NP_001230170.1 | 4 | 1082 | |||
NM_001243242.1 | NP_001230171.1 | 4 | 779 | |||
NM_001243243.1 | NP_001230172.1 | 3 | 681 | |||
NM_001243244.1 | NP_001230173.1 | 3 | 670 | |||
NM_030899.4 | NP_112161.3 | 4 | 1146 | |||
NM_145909.2 | NP_665916.1 | 8 | 1283 | |||
XM_005249295.1 | XP_005249352.1 | 4 | 1008 | |||
XM_005249296.4 | XP_005249353.1 | 5 | 5283 | |||
XM_011514807.2 | XP_011513109.1 | 6 | 5333 | |||
XM_011514808.2 | XP_011513110.1 | 4 | 5129 | |||
XM_011514809.1 | XP_011513111.1 | 4 | 993 | |||
XM_011514811.2 | XP_011513113.1 | 4 | 1508 | |||
XM_011514812.2 | XP_011513114.1 | 4 | 1034 | |||
XM_011514813.2 | XP_011513115.1 | 4 | 1497 | |||
XM_017011196.1 | XP_016866685.1 | 6 | 5421 | |||
GenBank mRNA | AF513019.1 | - | 8 | 1283 | ||
AK027060.1 | - | 1 | 172 | |||
AK057702.1 | - | 8 | 1283 | |||
AK095261.1 | - | 8 | 1661 | |||
AK225382.1 | - | 1 | 175 | |||
AL833416.1 | - | 3 | 441 | |||
AL833617.1 | - | 4 | 1158 | |||
BC008490.1 | - | 4 | 920 | 4248596 | ||
BC065241.1 | - | 3 | 451 | 6016003 | ||
DA879276.1 | - | 4 | 538 |