Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022451.10 | NP_071896.8 | 10 | 1376 | ||
XM_017016544.1 | XP_016872033.1 | 8 | 1429 | |||
GenBank mRNA | AB077992.1 | - | 10 | 1263 | ||
AK022882.1 | - | 2 | 287 | |||
AK091246.1 | - | 10 | 1333 | |||
AK292520.1 | - | 10 | 1358 | |||
AK302028.1 | - | 4 | 461 | |||
BC049850.1 | - | 10 | 1326 | 5549517 |