Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022458.3 | NP_071903.2 | 3, 4 | 402 | ||
XM_005249555.2 | XP_005249612.1 | 3, 4 | 377 | |||
XM_005249558.2 | XP_005249615.1 | 3, 4 | 377 | |||
XM_017012507.1 | XP_016867996.1 | 3, 4 | 377 | |||
XM_017012509.1 | XP_016867998.1 | 3, 4 | 377 | |||
XM_017012514.1 | XP_016868003.1 | 4, 5 | 471 | |||
XM_017012515.1 | XP_016868004.1 | 3, 4 | 377 | |||
XM_017012517.1 | XP_016868006.1 | 3, 4 | 377 | |||
XM_017012518.1 | XP_016868007.1 | 4, 5 | 475 | |||
GenBank mRNA | AF402318.1 | - | 3, 4 | 213 | ||
AK021727.1 | - | 3, 4 | 373 | |||
AK291037.1 | - | 3, 4 | 373 | |||
AK299753.1 | - | 2, 3 | 300 | |||
AL834394.1 | - | 3, 4 | 181 | |||
BC017663.1 | - | 3, 4 | 341 | 3883022 | ||
BC063649.1 | - | 4, 5 | 471 | |||
DA238712.1 | - | 3, 4 | 402 |