Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022458.3 | NP_071903.2 | 16 | 1580 | ||
XM_017012507.1 | XP_016867996.1 | 17 | 1678 | |||
XM_017012508.1 | XP_016867997.1 | 17 | 1636 | |||
XM_017012510.1 | XP_016867999.1 | 15 | 1282 | |||
XM_017012511.1 | XP_016868000.1 | 15 | 1411 | |||
XM_017012512.1 | XP_016868001.1 | 15 | 1342 | |||
XM_017012513.1 | XP_016868002.1 | 16 | 1360 | |||
XM_017012514.1 | XP_016868003.1 | 17 | 1649 | |||
XM_017012517.1 | XP_016868006.1 | 17 | 1633 | |||
XM_017012518.1 | XP_016868007.1 | 18 | 1731 | |||
GenBank mRNA | AF348513.1 | - | 3 | 300 | ||
AF402318.1 | - | 16 | 1391 | |||
AK026940.1 | - | 3 | 306 | |||
AK291037.1 | - | 16 | 1551 | |||
AL834394.1 | - | 17 | 1482 | |||
BC017663.1 | - | 16 | 1519 | 3883022 | ||
BC063649.1 | - | 18 | 1727 |