Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022460.3 | NP_071905.3 | 3 | 271 | ||
XM_017004696.1 | XP_016860185.1 | 3 | 274 | |||
XM_017004697.1 | XP_016860186.1 | 3 | 274 | |||
XM_017004698.1 | XP_016860187.1 | 3 | 274 | |||
XM_017004699.1 | XP_016860188.1 | 3 | 326 | |||
XM_017004700.1 | XP_016860189.1 | 3 | 370 | |||
XM_017004701.1 | XP_016860190.1 | 3 | 274 | |||
GenBank mRNA | AK024311.1 | - | 3 | 264 | ||
AK314383.1 | - | 3 | 267 | |||
BC050636.1 | - | 3 | 251 | 5193161 | ||
BX354596.2 | - | 3 | 302 |