Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022460.3 | NP_071905.3 | 7 | 1063 | ||
XM_017004700.1 | XP_016860189.1 | 7 | 1162 | |||
GenBank mRNA | AK314383.1 | - | 7 | 1059 | ||
BC050636.1 | - | 7 | 1043 | 5193161 | ||
BG753647.1 | - | 4 | 453 |