Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037633.1 | NP_001032722.1 | 7 | 768 | ||
NM_022464.4 | NP_071909.1 | 6 | 680 | |||
XM_011543570.2 | XP_011541872.1 | 8 | 805 | |||
GenBank mRNA | AF547994.1 | - | 6 | 599 | ||
AJ299442.1 | - | 6 | 658 | |||
AK074624.1 | - | 6 | 672 | |||
AK075177.1 | - | 6 | 609 | |||
AY358950.1 | - | 6 | 626 | |||
BC011568.1 | - | 6 | 648 | 4640182 | ||
KU178743.1 | - | 5 | 562 |