Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127895.1 | NP_001121367.1 | 5 | 1725 | ||
NM_001127896.1 | NP_001121368.1 | 4 | 1636 | |||
NM_022467.3 | NP_071912.2 | 4 | 1978 | |||
XM_011527222.1 | XP_011525524.1 | 4 | 1959 | |||
XM_011527224.1 | XP_011525526.1 | 5 | 2093 | |||
XM_017027143.1 | XP_016882632.1 | 4 | 1629 | |||
GenBank mRNA | AB047801.1 | - | 4 | 1492 | ||
AF300612.1 | - | 4 | 1439 | |||
AF305781.1 | - | 4 | 1535 | |||
BC011380.1 | - | 4 | 1588 | 3874578 | ||
BC014250.2 | - | 4 | 1632 | 4635405 | ||
BC018723.1 | - | 4 | 1978 | 4137487 |