Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022468.4 | NP_071913.1 | 9, 10 | 1654 | ||
XM_011522602.2 | XP_011520904.1 | 9, 10 | 1391 | |||
XM_011522604.2 | XP_011520906.1 | 8, 9 | 1505 | |||
XM_011522605.2 | XP_011520907.1 | 6, 7 | 845 | |||
GenBank mRNA | AB042328.1 | - | 9, 10 | 1586 | ||
AF145442.2 | - | 9, 10 | 1441 | |||
AF185270.1 | - | 9, 10 | 1441 | |||
AJ239053.1 | - | 9, 10 | 1598 | |||
AJ272137.1 | - | 9, 10 | 1654 |