Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284306.1 | NP_001271235.1 | 4 | 1107 | ||
NM_001284307.1 | NP_001271236.1 | 6 | 1122 | |||
NM_022473.2 | NP_071918.1 | 5 | 3550 | |||
XM_005254591.2 | XP_005254648.1 | 8 | 3457 | |||
XM_005254592.2 | XP_005254649.1 | 8 | 3457 | |||
XM_017022483.1 | XP_016877972.1 | 7 | 3232 | |||
XM_017022484.1 | XP_016877973.1 | 5 | 2977 | |||
XM_017022485.1 | XP_016877974.1 | 6 | 1117 | |||
GenBank mRNA | AF205632.1 | - | 5 | 3523 | ||
AK127741.1 | - | 3 | 350 | |||
AK226132.1 | - | 6 | 1030 | |||
AK302735.1 | - | 7 | 1004 | |||
AL831983.1 | - | 4 | 1053 | |||
AL832014.1 | - | 4 | 1331 | |||
AL832352.1 | - | 4 | 1550 | |||
AL832367.1 | - | 3 | 262 | |||
AL832455.2 | - | 4 | 1464 | |||
AL833301.1 | - | 4 | 1561 | |||
BX648629.1 | - | 4 | 2655 |