Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022473.2 | NP_071918.1 | 2 | 2188 | ||
XM_005254591.2 | XP_005254648.1 | 5 | 2095 | |||
XM_005254592.2 | XP_005254649.1 | 5 | 2095 | |||
XM_017022483.1 | XP_016877972.1 | 4 | 1870 | |||
XM_017022484.1 | XP_016877973.1 | 2 | 1615 | |||
GenBank mRNA | AF205632.1 | - | 2 | 2161 | ||
AK057410.1 | - | 1 | 1169 | |||
AK091929.1 | - | 3 | 1924 | |||
AK294529.1 | - | 2 | 1918 | |||
AL832352.1 | - | 1 | 188 | |||
AL832455.2 | - | 1 | 102 | |||
AL833301.1 | - | 1 | 199 | |||
BX648629.1 | - | 1 | 1293 |