Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000573.3 | NP_000564.2 | 31, 32 | 5365 | ||
NM_000651.4 | NP_000642.3 | 39, 40 | 6715 | |||
XM_006711166.2 | XP_006711229.1 | 39, 40 | 6691 | |||
XM_011509205.1 | XP_011507507.1 | 39, 40 | 6691 | |||
GenBank mRNA | X05309.1 | - | 22, 23 | 3721 | ||
Y00816.1 | - | 5252 |