Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282766.1 | NP_001269695.1 | 11, 12 | 1179 | ||
NM_022490.2 | NP_071935.1 | 10, 11 | 1085 | |||
XM_005251547.2 | XP_005251604.1 | 9, 10 | 1330 | |||
GenBank mRNA | AK023452.1 | - | 10, 11 | 1072 | ||
AK024032.1 | - | 10, 11 | 1061 | |||
AK091294.1 | - | 9, 10 | 1446 | |||
AK225050.1 | - | 10, 11 | 1058 | |||
AK301347.1 | - | 8, 9 | 762 | |||
AK302345.1 | - | 9, 10 | 941 | |||
AK303167.1 | - | 11, 12 | 1166 | |||
BC001337.1 | - | 10, 11 | 1052 | 3461710 | ||
BC014331.1 | - | 10, 11 | 1086 | 4042929 | ||
CR457313.1 | - | 10, 11 | 972 |