Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022491.2 | NP_071936.2 | 5, 6 | 509 | ||
XM_011538671.2 | XP_011536973.1 | 6, 7 | 1586 | |||
XM_017019817.1 | XP_016875306.1 | 4, 5 | 393 | |||
XM_017019818.1 | XP_016875307.1 | 7, 8 | 741 | |||
XM_017019819.1 | XP_016875308.1 | 6, 7 | 603 | |||
XM_017019820.1 | XP_016875309.1 | 7, 8 | 635 | |||
GenBank mRNA | AK024460.1 | - | 5, 6 | 492 | ||
BC098404.1 | - | 5, 6 | 532 | 30332266 | ||
BX647687.1 | - | 6, 7 | 467 |