Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022495.5 | NP_071940.4 | 5, 6 | 1400 | ||
XM_005268009.2 | XP_005268066.1 | 6, 7 | 2777 | |||
GenBank mRNA | AK095489.1 | - | 5, 6 | 1384 | ||
BX648440.1 | - | 1, 2 | 124 |