Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172810.1 | NP_001166281.1 | 4 | 496 | ||
NM_023036.4 | NP_075462.3 | 4 | 499 | |||
XM_011525125.2 | XP_011523427.1 | 4 | 516 | |||
GenBank mRNA | AF250288.1 | - | 4 | 456 | ||
AJ295276.1 | - | 3 | 695 | |||
BC039582.1 | - | 4 | 486 | 5752153 | ||
BI462861.1 | - | 4 | 495 | |||
BI464166.1 | - | 4 | 485 |