Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079533.1 | NP_001073001.1 | 2 | 347 | ||
NM_001079534.1 | NP_001073002.1 | 2 | 347 | |||
NM_001079535.1 | NP_001073003.1 | 1 | 448 | |||
NM_001288819.1 | NP_001275748.1 | 3 | 801 | |||
NM_001288820.1 | NP_001275749.1 | 2 | 347 | |||
NM_030594.4 | NP_085097.3 | 3 | 493 | |||
GenBank mRNA | AF329402.1 | - | 3 | 664 | ||
AF329403.1 | - | 2 | 330 | |||
AK023265.1 | - | 2 | 287 | |||
AK294280.1 | - | 2 | 373 | |||
AK300105.1 | - | 3 | 801 | |||
AK301728.1 | - | 2 | 308 | |||
BC035348.1 | - | 1 | 375 | 5195793 | ||
BC050629.1 | - | 3 | 454 | 6047179 | ||
BX649159.1 | - | 4 | 2233 | |||
DA340952.1 | - | 1 | 339 | |||
DA705313.1 | - | 2 | 347 |