Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193480.1 | NP_001180409.1 | 1 | 142 | ||
NM_022568.3 | NP_072090.1 | 1 | 142 | |||
NM_170771.2 | NP_739577.1 | 1 | 142 | |||
GenBank mRNA | AF303134.1 | - | 1 | 133 | ||
AK074266.1 | - | 1 | 135 | |||
AK222848.1 | - | 1 | 135 | |||
AK290784.1 | - | 1 | 89 | |||
AK298325.1 | - | 1 | 104 | |||
BC113862.1 | - | 1 | 112 | 40033493 | ||
BC114473.1 | - | 1 | 109 | 40033488 | ||
DA635162.1 | - | 1 | 142 |