Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022571.5 | NP_072093.2 | 1 | 1554 | ||
XM_017021596.1 | XP_016877085.1 | 1 | 1569 | |||
XM_017021597.1 | XP_016877086.1 | 1 | 1569 | |||
XM_017021598.1 | XP_016877087.1 | 1 | 1569 | |||
XM_017021599.1 | XP_016877088.1 | 1 | 1569 | |||
GenBank mRNA | AY288418.1 | - | 1 | 1439 | ||
BC032831.2 | - | 1 | 1554 | 4825908 | ||
BX094623.1 | - | 1 | 433 |
Set Membership: |
G-protein coupled receptor |