Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIGYF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022574.4 | NP_072096.2 | 17 | 3050 | ||
XM_005250532.1 | XP_005250589.1 | 19 | 2831 | |||
XM_011516472.1 | XP_011514774.1 | 19 | 2831 | |||
XM_011516477.1 | XP_011514779.1 | 19 | 2831 | |||
XM_017012526.1 | XP_016868015.1 | 19 | 2831 | |||
XM_017012527.1 | XP_016868016.1 | 21 | 2729 | |||
XM_017012528.1 | XP_016868017.1 | 20 | 2341 | |||
XM_017012529.1 | XP_016868018.1 | 20 | 2348 | |||
XM_017012530.1 | XP_016868019.1 | 20 | 2602 | |||
XM_017012531.1 | XP_016868020.1 | 15 | 1854 | |||
GenBank mRNA | AY176044.1 | - | 17 | 3251 | ||
BC018563.1 | - | 1 | 24 | |||
BC129991.1 | - | 11 | 1198 | |||
BQ069946.1 | - | 5 | 485 |