Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022575.3 | NP_072097.2 | 22 | 2291 | ||
NM_080413.2 | NP_536338.1 | 18 | 1859 | |||
GenBank mRNA | AF308801.1 | - | 22 | 2227 | ||
AK055787.1 | - | 18 | 1829 | |||
AL834401.1 | - | 18 | 1779 | |||
BC012422.1 | - | 10 | 942 | |||
BC021291.2 | - | 8 | 810 | |||
BC064406.1 | - | 6 | 547 | |||
BC073959.1 | - | 10 | 1361 | 6182570 | ||
BC126431.1 | - | 7 | 643 | |||
BM468030.1 | - | 3 | 264 |