Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022659.3 | NP_073150.2 | 3 | 588 | ||
GenBank mRNA | AK001144.1 | - | 3 | 641 | ||
AK304372.1 | - | 2 | 236 | |||
AL701879.1 | - | 3 | 237 | |||
BC137106.1 | - | 3 | 588 | |||
BC137107.1 | - | 3 | 588 |