Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033055.2 | NP_149044.2 | 11 | 1134 | ||
XM_005271118.1 | XP_005271175.1 | 12 | 1410 | |||
XM_011541957.1 | XP_011540259.1 | 10 | 1175 | |||
XM_017002084.1 | XP_016857573.1 | 12 | 1377 | |||
GenBank mRNA | AF427492.1 | - | 11 | 1134 | ||
AK057172.1 | - | 11 | 1270 | |||
AK075208.1 | - | 5 | 335 | |||
AK096669.1 | - | 8 | 785 | |||
AL137576.1 | - | 3 | 228 | |||
BC064409.1 | - | 8 | 791 | |||
BC107768.1 | - | 8 | 883 |