Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004378.2 | NP_004369.1 | 3 | 433 | ||
GenBank mRNA | AB593091.1 | - | 3 | 407 | ||
AL110113.1 | - | 3 | 317 | |||
AU132726.1 | - | 3 | 433 | |||
CR457017.1 | - | 3 | 328 | |||
S74445.1 | - | 3 | 402 |