Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282949.2 | NP_001269878.1 | 2 | 278 | ||
NM_001282950.2 | NP_001269879.1 | 3 | 399 | |||
NM_001308381.1 | NP_001295310.1 | 3 | 399 | |||
NM_022730.3 | NP_073567.1 | 3 | 399 | |||
XM_006712693.1 | XP_006712756.1 | 3 | 456 | |||
XM_011511638.1 | XP_011509940.1 | 2 | 1839 | |||
XM_017004716.1 | XP_016860205.1 | 3 | 456 | |||
GenBank mRNA | AK022674.1 | - | 3 | 330 | ||
AK124133.1 | - | 3 | 382 | |||
AK126326.1 | - | 2 | 1586 | |||
AK222879.1 | - | 3 | 337 | |||
AK307486.1 | - | 2 | 216 | |||
BC091493.1 | - | 3 | 334 | 30387712 | ||
BX333333.2 | - | 3 | 337 | |||
DA754035.1 | - | 3 | 457 |