Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009606.3 | NP_001009606.3 | 2 | 475 | ||
XM_011522608.2 | XP_011520910.1 | 2 | 701 | |||
XM_011522609.2 | XP_011520911.1 | 2 | 2182 |