Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198978.1 | NP_001185907.1 | 8 | 745 | ||
NM_001198979.1 | NP_001185908.1 | 8 | 829 | |||
NM_001198980.1 | NP_001185909.1 | 8 | 760 | |||
NM_022733.2 | NP_073570.1 | 8 | 1187 | |||
XM_011541960.1 | XP_011540262.1 | 9 | 1194 | |||
XM_017002093.1 | XP_016857582.1 | 5 | 886 | |||
GenBank mRNA | AK294675.1 | - | 3 | 469 | ||
AK298702.1 | - | 8 | 725 | |||
AK303975.1 | - | 8 | 733 | |||
AK308280.1 | - | 8 | 830 | |||
AK313105.1 | - | 8 | 1134 | |||
AK316218.1 | - | 8 | 760 | |||
AL133206.1 | - | 8 | 701 | |||
AL137764.1 | - | 8 | 714 | |||
BC014512.1 | - | 6 | 526 | |||
BC021133.1 | - | 8 | 837 | 4855320 |