Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199723.1 | NP_001186652.1 | 3 | 398 | ||
NM_001878.3 | NP_001869.1 | 2 | 411 | |||
XM_017000343.1 | XP_016855832.1 | 5 | 5825 | |||
GenBank mRNA | AB593017.1 | - | 2 | 327 | ||
AK312007.1 | - | 2 | 327 | |||
BC001109.2 | - | 2 | 313 | 2988143 | ||
BP231157.1 | - | 2 | 411 | |||
BT019827.1 | - | 2 | 190 | |||
CR450357.1 | - | 2 | 190 | |||
M68867.1 | - | 2 | 288 |