Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167903.1 | NP_001161375.1 | 7 | 780 | ||
NM_001289406.1 | NP_001276335.1 | 7 | 784 | |||
NM_001289407.1 | NP_001276336.1 | 8 | 866 | |||
NM_022736.2 | NP_073573.2 | 8 | 897 | |||
NR_110328.1 | 7 | 784 | ||||
XM_006713730.2 | XP_006713793.1 | 7 | 715 | |||
GenBank mRNA | AB014732.1 | - | 8 | 729 | ||
AF351617.2 | - | 8 | 593 | |||
AK024215.1 | - | 8 | 700 | |||
AK294628.1 | - | 7 | 657 | |||
AK297034.1 | - | 488 | ||||
AK297593.1 | - | 8 | 897 | |||
AK300497.1 | - | 7 | 583 | |||
AK301680.1 | - | 8 | 636 | |||
BC030542.1 | - | 7 | 653 | 4915325 | ||
BC042197.1 | - | 8 | 709 | 5737259 |