Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000755.3 | NP_000746.2 | 6 | 990 | ||
NM_001257363.1 | NP_001244292.1 | 7 | 1089 | |||
XM_005251708.3 | XP_005251765.1 | 8 | 1152 | |||
XM_017014274.1 | XP_016869763.1 | 7 | 1157 | |||
XM_017014275.1 | XP_016869764.1 | 6 | 810 | |||
GenBank mRNA | AK313931.1 | - | 7 | 1001 | ||
BC000723.2 | - | 6 | 929 | 3504259 | ||
BT006801.1 | - | 6 | 708 | |||
X78706.1 | - | 6 | 703 |