Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022744.3 | NP_073581.2 | 1 | 194 | ||
XM_017023570.1 | XP_016879059.1 | 1 | 164 | |||
GenBank mRNA | AK023930.1 | - | 1 | 160 | ||
AK222913.1 | - | 1 | 160 | |||
AK296166.1 | - | 1 | 159 | |||
AK296491.1 | - | 1 | 161 | |||
BC009308.2 | - | 1 | 154 | 4301507 | ||
DA742232.1 | - | 1 | 194 |