Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000755.3 | NP_000746.2 | 7 | 1171 | ||
NM_001257363.1 | NP_001244292.1 | 8 | 1270 | |||
XM_005251708.3 | XP_005251765.1 | 9 | 1333 | |||
XM_017014274.1 | XP_016869763.1 | 8 | 1338 | |||
XM_017014275.1 | XP_016869764.1 | 7 | 991 | |||
GenBank mRNA | AK313931.1 | - | 8 | 1182 | ||
X78706.1 | - | 7 | 884 |