Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022750.3 | NP_073587.1 | 10 | 2388 | ||
NR_130117.1 | 9 | 2246 | ||||
GenBank mRNA | AK026346.1 | - | 4 | 228 | ||
AL136766.1 | - | 10 | 2010 | |||
AL137255.1 | - | 10 | 1207 | |||
BC081541.1 | - | 10 | 2388 | 5296560 |