Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022752.5 | NP_073589.4 | 2 | 2608 | ||
XM_005259159.2 | XP_005259216.1 | 1 | 2471 | |||
XM_011527228.2 | XP_011525530.1 | 2 | 2801 | |||
XM_011527229.2 | XP_011525531.1 | 2 | 2524 | |||
XM_011527230.2 | XP_011525532.1 | 2 | 2476 | |||
XM_017027149.1 | XP_016882638.1 | 2 | 2944 | |||
GenBank mRNA | AF447880.1 | - | 2 | 2553 | ||
AK025712.1 | - | 1 | 1716 | |||
AK027895.1 | - | 2 | 2509 | |||
AK131369.1 | - | 2 | 2944 | |||
AK131418.1 | - | 2 | 3094 | |||
AK225342.1 | - | 1 | 1716 | |||
AL137516.1 | - | 1 | 1685 | |||
BC001184.1 | - | 2 | 2497 | 3502564 | ||
BC071962.1 | - | 1 | 1261 |