Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318246.1 | NP_001305175.1 | 11 | 1271 | ||
NM_194071.3 | NP_919047.2 | 11 | 1684 | |||
XM_005250541.4 | XP_005250598.1 | 11 | 2063 | |||
XM_017012540.1 | XP_016868029.1 | 12 | 3086 | |||
XM_017012541.1 | XP_016868030.1 | 12 | 1649 | |||
GenBank mRNA | AJ549092.1 | - | 11 | 1637 | ||
AJ549387.1 | - | 11 | 1637 | |||
AK293048.1 | - | 11 | 1616 | |||
BC110813.1 | - | 11 | 1620 | 3923538 | ||
BX649143.1 | - | 11 | 1276 | |||
CR749563.1 | - | 10 | 1364 |