Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100PBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256121.1 | NP_001243050.1 | 3 | 667 | ||
NM_022753.3 | NP_073590.2 | 3 | 681 | |||
NR_135106.1 | 2 | 509 | ||||
NR_135107.1 | 2 | 509 | ||||
XM_011541961.2 | XP_011540263.1 | 3 | 616 | |||
XM_011541962.2 | XP_011540264.1 | 3 | 725 | |||
XM_011541963.1 | XP_011540265.1 | 3 | 681 | |||
XM_017002098.1 | XP_016857587.1 | 3 | 681 | |||
GenBank mRNA | AK090711.1 | - | 3 | 667 | ||
BC015175.1 | - | 3 | 540 | 3904501 | ||
BX647667.1 | - | 3 | 608 | |||
BX647916.1 | - | 3 | 645 | |||
KU178750.1 | - | 1 | 390 | |||
KU178751.1 | - | 1 | 390 |