Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100PBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256121.1 | NP_001243050.1 | 3 | 1004 | ||
NM_022753.3 | NP_073590.2 | 3 | 1018 | |||
NR_135106.1 | 2 | 846 | ||||
NR_135107.1 | 2 | 846 | ||||
XM_011541961.2 | XP_011540263.1 | 3 | 953 | |||
XM_011541962.2 | XP_011540264.1 | 3 | 1062 | |||
XM_011541963.1 | XP_011540265.1 | 3 | 1018 | |||
XM_017002098.1 | XP_016857587.1 | 3 | 1018 | |||
GenBank mRNA | AK022965.1 | - | 1 | 325 | ||
AK090711.1 | - | 3 | 1004 | |||
BC015175.1 | - | 3 | 877 | 3904501 | ||
BX647667.1 | - | 3 | 945 | |||
BX647916.1 | - | 3 | 982 | |||
KU178750.1 | - | 1 | 727 | |||
KU178751.1 | - | 1 | 727 |