Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308317.1 | NP_001295246.1 | 10 | 1652 | ||
NM_022757.4 | NP_073594.4 | 10 | 1585 | |||
XM_005247710.4 | XP_005247767.1 | 11 | 1708 | |||
XM_005247711.4 | XP_005247768.1 | 11 | 1233 | |||
XM_005247713.2 | XP_005247770.1 | 8 | 1482 | |||
XM_005247714.2 | XP_005247771.1 | 7 | 1361 | |||
XM_005247715.4 | XP_005247772.1 | 11 | 1711 | |||
XM_006713731.3 | XP_006713794.1 | 11 | 1710 | |||
XM_006713733.3 | XP_006713796.1 | 11 | 1235 | |||
XM_011513081.2 | XP_011511383.1 | 11 | 1711 | |||
XM_017007056.1 | XP_016862545.1 | 7 | 5237 | |||
XM_017007057.1 | XP_016862546.1 | 8 | 1276 | |||
XM_017007058.1 | XP_016862547.1 | 9 | 1389 | |||
XM_017007059.1 | XP_016862548.1 | 7 | 1284 | |||
XM_017007060.1 | XP_016862549.1 | 7 | 1351 | |||
XM_017007061.1 | XP_016862550.1 | 7 | 1361 | |||
GenBank mRNA | AK027607.1 | - | 7 | 684 | ||
AK123060.1 | - | 8 | 2141 | |||
AK294979.1 | - | 11 | 1560 | |||
AK295091.1 | - | 10 | 1555 | |||
AK307847.1 | - | 10 | 1405 | |||
AL122079.1 | - | 11 | ||||
AL833312.1 | - | 8 | 2280 | |||
BC040285.1 | - | 10 | 1516 | 4825732 | ||
BC042613.1 | - | 7 | 4179 | 4822585 | ||
BX537652.1 | - | 7 | 5612 |