Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENGASE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042573.2 | NP_001036038.1 | 3 | 260 | ||
XM_006722019.3 | XP_006722082.1 | 3 | 385 | |||
XM_017024961.1 | XP_016880450.1 | 2 | 102 | |||
GenBank mRNA | AF512564.1 | - | 3 | 252 | ||
AK301115.1 | - | 3 | 258 |