Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022766.5 | NP_073603.2 | 13 | 2477 | ||
XM_017028909.1 | XP_016884398.1 | 13 | 2274 | |||
GenBank mRNA | AB051433.1 | - | 12 | 2203 | ||
AB079066.1 | - | 13 | 2493 | |||
AK026892.1 | - | 1 | 635 | |||
AL832794.1 | - | 3 | 1054 | |||
BC004278.1 | - | 3 | 1054 | |||
BC108712.1 | - | 2 | 979 | |||
CR457365.1 | - | 1 | 223 |