Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126129.1 | NP_001119601.1 | 4 | 929 | ||
NM_001126130.1 | NP_001119602.1 | 2 | 578 | |||
NM_022770.3 | NP_073607.2 | 3 | 812 | |||
GenBank mRNA | AK023974.1 | - | 2 | 484 | ||
AK127454.1 | - | 1 | 1741 | |||
AK315622.1 | - | 3 | 763 | |||
AL137379.1 | - | 1 | 148 | |||
BC005879.1 | - | 3 | 687 | 3529058 | ||
BC014437.1 | - | 3 | 706 | 4870865 | ||
CR457283.1 | - | 2 | 370 | |||
KU178752.1 | - | 3 | 604 | |||
KU178753.1 | - | 4 | 721 | |||
KU178754.1 | - | 4 | 663 | |||
KU178755.1 | - | 2 | 370 |