Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146213.1 | NP_001139685.2 | 7 | 763 | ||
NM_001146214.1 | NP_001139686.1 | 8 | 855 | |||
NM_022771.4 | NP_073608.4 | 8 | 814 | |||
NR_027449.1 | 6 | 552 | ||||
XM_006719564.2 | XP_006719627.1 | 8 | 796 | |||
XM_006719565.3 | XP_006719628.1 | 8 | 734 | |||
XM_006719566.2 | XP_006719629.1 | 9 | 945 | |||
XM_006719567.2 | XP_006719630.1 | 7 | 631 | |||
XM_011538679.1 | XP_011536981.1 | 10 | 871 | |||
XM_011538680.1 | XP_011536982.1 | 8 | 829 | |||
XM_011538681.2 | XP_011536983.1 | 8 | 778 | |||
XM_011538682.1 | XP_011536984.1 | 7 | 633 | |||
XM_017019824.1 | XP_016875313.1 | 9 | 831 | |||
XM_017019825.1 | XP_016875314.1 | 8 | 850 | |||
XM_017019826.1 | XP_016875315.1 | 8 | 780 | |||
XM_017019827.1 | XP_016875316.1 | 5 | 454 | |||
XM_017019828.1 | XP_016875317.1 | 6 | 572 | |||
GenBank mRNA | AB449901.1 | - | 7 | 688 | ||
AK022147.1 | - | 7 | 711 | |||
AK225221.1 | - | 7 | 692 | |||
AK225747.1 | - | 7 | 709 | |||
AK292306.1 | - | 6 | 483 | |||
AK297626.1 | - | 8 | 803 | |||
AK307922.1 | - | 8 | 780 | |||
AL833639.1 | - | 6 | 491 | |||
BC028352.2 | - | 8 | 762 | 4827246 |