Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022772.3 | NP_073609.2 | 19 | 2050 | ||
XM_017018130.1 | XP_016873619.1 | 20 | 2267 | |||
XM_017018131.1 | XP_016873620.1 | 19 | 2138 | |||
XM_017018132.1 | XP_016873621.1 | 20 | 2108 | |||
GenBank mRNA | AF318331.1 | - | 7 | 1221 | ||
AK025588.1 | - | 19 | 1968 | |||
AK025824.1 | - | 19 | 1968 | |||
AK027765.1 | - | 7 | 862 | |||
AK094539.1 | - | 11 | 1146 | |||
AK122903.1 | - | 16 | 2246 | |||
AK122984.1 | - | 4 | 516 | |||
AK222903.1 | - | 19 | 1963 | |||
AK225311.1 | - | 19 | 1968 | |||
AY074929.1 | - | 19 | 2075 | |||
BC080636.1 | - | 19 | 2054 | 6572993 | ||
BC093878.1 | - | 18 | 1843 | 7939723 | ||
BC101481.1 | - | 18 | 1843 | 8068987 | ||
BC143242.1 | - | 18 | 1891 | 9051743 |