Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022772.3 | NP_073609.2 | 16 | 1739 | ||
XM_017018130.1 | XP_016873619.1 | 17 | 1956 | |||
XM_017018131.1 | XP_016873620.1 | 16 | 1827 | |||
XM_017018132.1 | XP_016873621.1 | 17 | 1797 | |||
GenBank mRNA | AF318331.1 | - | 4 | 908 | ||
AK025588.1 | - | 16 | 1657 | |||
AK025824.1 | - | 16 | 1657 | |||
AK027765.1 | - | 4 | 551 | |||
AK094539.1 | - | 8 | 835 | |||
AK122903.1 | - | 13 | 1935 | |||
AK122984.1 | - | 1 | 205 | |||
AK222903.1 | - | 16 | 1652 | |||
AK225311.1 | - | 16 | 1657 | |||
AY074929.1 | - | 16 | 1764 | |||
BC080636.1 | - | 16 | 1743 | 6572993 | ||
BC093878.1 | - | 15 | 1532 | 7939723 | ||
BC101481.1 | - | 15 | 1532 | 8068987 | ||
BC143242.1 | - | 15 | 1580 | 9051743 |