Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022773.2 | NP_073610.2 | 4, 5 | 670 | ||
NR_033645.1 | 5, 6 | 799 | ||||
NR_033646.1 | 6, 7 | 977 | ||||
XM_006720928.2 | XP_006720991.1 | 4, 5 | 943 | |||
XM_006720929.3 | XP_006720992.1 | 5, 6 | 824 | |||
XM_011522614.2 | XP_011520916.1 | 4, 5 | 668 | |||
XM_011522615.1 | XP_011520917.1 | 4, 5 | 668 | |||
XM_011522617.2 | XP_011520919.1 | 2, 3 | 319 | |||
XM_011522618.2 | XP_011520920.1 | 4, 5 | 668 | |||
XM_017023575.1 | XP_016879064.1 | 4, 5 | 668 | |||
XM_017023576.1 | XP_016879065.1 | 3, 4 | 434 | |||
XM_017023577.1 | XP_016879066.1 | 5, 6 | 1099 | |||
XM_017023579.1 | XP_016879068.1 | 3, 4 | 527 | |||
XM_017023580.1 | XP_016879069.1 | 6, 7 | 1249 | |||
XM_017023581.1 | XP_016879070.1 | 4, 5 | 668 | |||
GenBank mRNA | AB075873.1 | - | 4, 5 | 654 | ||
AK022743.1 | - | 4, 5 | 668 | |||
AK025955.1 | - | 4, 5 | 602 | |||
AK125201.1 | - | 5, 6 | 801 | |||
AK225065.1 | - | 4, 5 | 668 | |||
AK225406.1 | - | 4, 5 | 655 | |||
AK294329.1 | - | 6, 7 | 975 | |||
AK294932.1 | - | 4, 5 | 703 | |||
BC010738.1 | - | 3, 4 | 435 |