Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281517.2 | NP_001268446.1 | 6, 7 | 1376 | ||
NM_001319944.1 | NP_001306873.1 | 7, 8 | 1529 | |||
NM_022778.4 | NP_073615.2 | 7, 8 | 1529 | |||
XM_017002105.1 | XP_016857594.1 | 7, 8 | 1529 | |||
GenBank mRNA | AK024038.1 | - | 7, 8 | 1485 | ||
AK025653.1 | - | 1, 2 | 128 | |||
AK299312.1 | - | 6, 7 | 1329 | |||
AL133609.1 | - | 3, 4 | 370 | |||
AL137420.1 | - | 1, 2 | 127 | |||
BC064528.1 | - | 7, 8 | 1480 | 5585742 |